This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC61 SIRNA Assays ›
Gene Symbol:
Lrrc61
Gene Name:
leucine rich repeat containing 61
Gene Aliases:
RGD1561800
Chromosome Location:
Chr.4: 78185946 - 78206818 on Build Rnor_6.0
UniGene ID:
Rn.140918
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS365288
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc61
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109231.1 NP_001102701.1 3 661
XM_006236435.2 XP_006236497.1 2 1678
XM_006236436.2 XP_006236498.1 2 529
XM_006236437.2 XP_006236499.1 2 696
XM_006236438.2 XP_006236500.1 2 381

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0