This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP1R3A SIRNA Assays ›
Gene Symbol:
Ppp1r3a
Gene Name:
protein phosphatase 1, regulatory subunit 3A
Gene Aliases:
RGD1561312
Chromosome Location:
Chr.4: 41171486 - 41212117 on Build Rnor_6.0
UniGene ID:
Rn.29809
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS365188
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp1r3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109222.1 NP_001102692.1 4 2120
XM_008762717.1 XP_008760939.1 4 2162

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm