This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRPF38B SIRNA Assays ›
Gene Symbol:
Prpf38b
Gene Name:
pre-mRNA processing factor 38B
Gene Aliases:
-
Chromosome Location:
Chr.2: 211706245 - 211715271 on Build Rnor_6.0
UniGene ID:
Rn.7210
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS364816
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prpf38b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024305.2 NP_001019476.1 6 1857
XM_006233172.1 XP_006233234.1 7 1613
XM_008761421.1 XP_008759643.1 6 1908
GenBank mRNA BC079264.1 - 6 1806 7133560
DY314904.1 - 1 52
FQ210223.1 - 6 1835

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0