This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MSH3 SIRNA Assays ›
Gene Symbol:
Msh3
Gene Name:
mutS homolog 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 21790048 - 21931791 on Build Rnor_6.0
UniGene ID:
Rn.162551
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS364634
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Msh3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191957.1 NP_001178886.1 17 2293
XM_006231768.2 XP_006231830.1 17 2417
XM_006231769.2 XP_006231831.1 17 2221

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

meiotic mismatch repair
DNA repair
mismatch repair
reciprocal meiotic recombination
biological_process
somatic hypermutation of immunoglobulin genes
somatic recombination of immunoglobulin gene segments
maintenance of DNA repeat elements
negative regulation of DNA recombination
positive regulation of helicase activity
Y-form DNA binding
heteroduplex DNA loop binding
double-strand/single-strand DNA junction binding
molecular_function
DNA binding
damaged DNA binding
single-stranded DNA binding
ATP binding
DNA-dependent ATPase activity
centromeric DNA binding
enzyme binding
mismatched DNA binding
guanine/thymine mispair binding
dinucleotide insertion or deletion binding
single guanine insertion binding
dinucleotide repeat insertion binding
oxidized purine DNA binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0