siRNA Details
Gene Symbol: LOC498675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109113.1 | NP_001102583.1 | 4 | 605 |

Gene Symbol: LOC498675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109113.1 | NP_001102583.1 | 4 | 605 |