siRNA Details
Gene Symbol: Grxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191935.1 | NP_001178864.1 | 4 | 924 |

Gene Symbol: Grxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191935.1 | NP_001178864.1 | 4 | 924 |