This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPN13 SIRNA Assays ›
Gene Symbol:
Ptpn13
Gene Name:
protein tyrosine phosphatase, non-receptor type 13
Gene Aliases:
Ptp-bl, ptp
Chromosome Location:
Chr.14: 7688551 - 7863679 on Build Rnor_6.0
UniGene ID:
Rn.35546
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS363549
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptpn13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100789.1 NP_001094259.1 7 1019
XM_006250672.2 XP_006250734.1 7 1034
XM_006250674.2 XP_006250736.1 7 1034
XM_008770051.1 XP_008768273.1 7 1034
XM_008770052.1 XP_008768274.1 7 1034

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006264, NM_080684, NM_080683, NM_080685, AB208904, and others.
This siRNA also targets the following transcripts in Mouse: NM_011204, AK084198, BC023913, AK080313, D83966, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
phosphatidylinositol 3-kinase regulatory subunit binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0