This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM222B SIRNA Assays ›
Gene Symbol:
Fam222b
Gene Name:
family with sequence similarity 222, member B
Gene Aliases:
RGD1566149
Chromosome Location:
Chr.10: 65351245 - 65416869 on Build Rnor_6.0
UniGene ID:
Rn.19152
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS363133
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam222b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109038.1 NP_001102508.1 3 1796
XM_006246944.1 XP_006247006.1 3 1815
XM_008768094.1 XP_008766316.1 2 2293
XM_008768095.1 XP_008766317.1 3 2426
XM_008768096.1 XP_008766318.1 3 1703

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145430, AK140562, AK085728, BC017647, AK047860, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0