This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APOLD1 SIRNA Assays ›
Gene Symbol:
Apold1
Gene Name:
apolipoprotein L domain containing 1
Gene Aliases:
Verge
Chromosome Location:
Chr.4: 168752142 - 168755024 on Build Rnor_6.0
UniGene ID:
Rn.87679
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS362481
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Apold1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003403.1 NP_001003403.1 1 422
GenBank mRNA AY673969.1 - 1 422

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
response to hypoxia
lipid transport
cell differentiation
endothelial cell activation
lipoprotein metabolic process
regulation of endothelial cell differentiation
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0