This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other USP11 SIRNA Assays ›
Gene Symbol:
Usp11
Gene Name:
ubiquitin specific peptidase 11
Gene Aliases:
Uhx1
Chromosome Location:
Chr.X: 1687820 - 1703991 on Build Rnor_6.0
UniGene ID:
Rn.2492
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS362381
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008861.2 NP_001008861.2 19, 20 2540
GenBank mRNA BC090333.1 - 19, 20 2540 7190710
BC105613.1 - 15, 16 1953

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145628, AK169116, AK172230, BC005470, AK031767, and others.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header