siRNA Details
Gene Symbol: Marco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109011.1 | NP_001102481.1 | 1, 2 | 420 |

Gene Symbol: Marco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109011.1 | NP_001102481.1 | 1, 2 | 420 |