This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATP6V1E2 SIRNA Assays ›
Gene Symbol:
Atp6v1e2
Gene Name:
ATPase, H transporting, lysosomal V1 subunit E2
Gene Aliases:
-
Chromosome Location:
Chr.6: 10510370 - 10577704 on Build Rnor_6.0
UniGene ID:
Rn.161896
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS361579
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp6v1e2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108979.1 NP_001102449.1 2 402
XM_006239715.2 XP_006239777.1 2 448
XM_006239716.2 XP_006239778.1 2 849
XM_008764465.1 XP_008762687.1 2 425
XM_008764466.1 XP_008762688.1 2 464

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ATP hydrolysis coupled proton transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
proton-transporting ATPase activity, rotational mechanism

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0