This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARL11 SIRNA Assays ›
Gene Symbol:
Arl11
Gene Name:
ADP-ribosylation factor-like 11
Gene Aliases:
-
Chromosome Location:
Chr.15: 41639963 - 41645685 on Build Rnor_6.0
UniGene ID:
Rn.23090
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS360990
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arl11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013433.1 NP_001013451.1 2 303
XM_006252135.2 XP_006252197.1 3 1070
XM_008770766.1 XP_008768988.1 3 1065
GenBank mRNA BC091183.1 - 2 303 7365828

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
small GTPase mediated signal transduction
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0