This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other P4HA2 SIRNA Assays ›
Gene Symbol:
P4ha2
Gene Name:
prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases:
-
Chromosome Location:
Chr.10: 39435227 - 39464134 on Build Rnor_6.0
UniGene ID:
Rn.79891
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359775
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: P4ha2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108275.1 NP_001101745.1 9 1326
XM_006246300.1 XP_006246362.1 9 1295
XM_006246302.1 XP_006246364.1 9 1295
XM_008767691.1 XP_008765913.1 9 1323
GenBank mRNA FQ228519.1 - 9 1231

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011031, NM_001136076, U16163, BC018411, AK053504, and others.

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-proline hydroxylation to 4-hydroxy-L-proline
oxidation-reduction process
procollagen-proline 4-dioxygenase activity
iron ion binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
L-ascorbic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0