This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARMC8 SIRNA Assays ›
Gene Symbol:
Armc8
Gene Name:
armadillo repeat containing 8
Gene Aliases:
RGD1310477
Chromosome Location:
Chr.8: 107723359 - 107819203 on Build Rnor_6.0
UniGene ID:
Rn.8146
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359545
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Armc8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173354.2 NP_001166825.1 10 912
XM_006243622.2 XP_006243684.1 10 1149
XM_006243623.2 XP_006243685.1 10 1149
XM_006243624.2 XP_006243686.1 10 1149
XM_006243625.2 XP_006243687.1 9 988
GenBank mRNA BC168192.1 - 10 820
FQ211639.1 - 9 975

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001166138, NM_028768, AK004793, BC038866, AK078280, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0