This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADAMTS7 SIRNA Assays ›
Gene Symbol:
Adamts7
Gene Name:
ADAM metallopeptidase with thrombospondin type 1 motif, 7
Gene Aliases:
ADAMTS7B, COMPase
Chromosome Location:
Chr.8: 97535777 - 97575661 on Build Rnor_6.0
UniGene ID:
Rn.23850
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359529
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adamts7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047101.1 NP_001040566.1 10 2017
XM_006243523.1 XP_006243585.1 9 1675
XM_006243524.1 XP_006243586.1 9 1675
XM_006243525.1 XP_006243587.1 9 1675
XM_006243526.1 XP_006243588.1 9 1675
XM_006243527.2 XP_006243589.1 9 3942
XM_008766447.1 XP_008764669.1 9 2050
GenBank mRNA AY257482.1 - 10 2017
AY327121.1 - 9 1435

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
negative regulation of chondrocyte differentiation
proteolysis involved in cellular protein catabolic process
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to BMP stimulus
metalloendopeptidase activity
metallopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0