This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BRD1 SIRNA Assays ›
Gene Symbol:
Brd1
Gene Name:
bromodomain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 129676119 - 129724303 on Build Rnor_6.0
UniGene ID:
Rn.9133
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS359431
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Brd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108103.1 NP_001101573.1 13 3150
XM_006242194.2 XP_006242256.1 13 3539
XM_006242196.2 XP_006242258.1 13 3146

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033274, AK220233, BC130273, BC057182, BC049267, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to immobilization stress
histone H3 acetylation
response to electrical stimulus
zinc ion binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0