siRNA Details
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107885.1 | NP_001101355.1 | 6 | 702 | ||
XM_006237248.1 | XP_006237310.1 | 6 | 720 |

Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107885.1 | NP_001101355.1 | 6 | 702 | ||
XM_006237248.1 | XP_006237310.1 | 6 | 720 |