This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SULF2 SIRNA Assays ›
Gene Symbol:
Sulf2
Gene Name:
sulfatase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 162791349 - 162872986 on Build Rnor_6.0
UniGene ID:
Rn.4228
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS358874
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sulf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034927.1 NP_001030099.1 15 2705
XM_006235595.2 XP_006235657.1 15 2772
XM_006235596.2 XP_006235658.1 15 2303
XM_008762506.1 XP_008760728.1 15 2353
GenBank mRNA AY742216.1 - 15 2705
BC091377.1 - 5 484
BC166410.1 - 15 2184 9026145

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
chondrocyte development
glomerular filtration
positive regulation of vascular endothelial growth factor production
esophagus smooth muscle contraction
positive regulation of Wnt signaling pathway
heparan sulfate proteoglycan metabolic process
glomerular basement membrane development
glial cell-derived neurotrophic factor receptor signaling pathway
negative regulation of fibroblast growth factor receptor signaling pathway
embryonic skeletal system development
cartilage development
bone development
innervation
arylsulfatase activity
calcium ion binding
N-acetylglucosamine-6-sulfatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0