This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other JADE1 SIRNA Assays ›
Gene Symbol:
Jade1
Gene Name:
jade family PHD finger 1
Gene Aliases:
Phf17
Chromosome Location:
Chr.2: 128454158 - 128514298 on Build Rnor_6.0
UniGene ID:
Rn.90858
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS358667
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Jade1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107670.1 NP_001101140.1 6 783
XM_006232301.2 XP_006232363.1 6 1057
XM_006232302.2 XP_006232364.1 6 866
XM_006232303.2 XP_006232365.1 6 985
XM_006232304.2 XP_006232366.1 6 746
XM_008760928.1 XP_008759150.1 6 1758
XM_008760929.1 XP_008759151.1 6 753

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

histone H3 acetylation
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K12 acetylation
histone H4-K16 acetylation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of G1/S transition of mitotic cell cycle
RNA polymerase II transcription coactivator activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header