This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR318 SIRNA Assays ›
Gene Symbol:
Olr318 Olr1874
Gene Name:
olfactory receptor 318
olfactory receptor 1874
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 228262242 - 228263198 on Build Rnor_6.0
Chromosome Location:
Chr.1: 228225398 - 228226959 on Build Rnor_6.0
UniGene ID:
Rn.141711
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS358508
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr318
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000505.1 NP_001000505.1 1 521
Gene Symbol: Olr1874
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728868.4 XP_002728914.2 1 1126

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001011524, NM_146936, BC148189, BC153816, BC150977.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0