This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SH3BP2 SIRNA Assays ›
Gene Symbol:
Sh3bp2
Gene Name:
SH3-domain binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 81435445 - 81473002 on Build Rnor_6.0
UniGene ID:
Rn.212791
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS357863
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sh3bp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100684.1 NP_001094154.1 8 1129
XM_006251247.2 XP_006251309.1 8 2168
XM_006251248.2 XP_006251310.1 8 2006
XM_006251249.2 XP_006251311.1 8 2220
XM_006251251.2 XP_006251313.1 8 1078
XM_006251252.2 XP_006251314.1 8 4706
XM_006251253.2 XP_006251315.1 8 1942
XM_008770284.1 XP_008768506.1 9 5673
XM_008770285.1 XP_008768507.1 8 1335
GenBank mRNA FQ210321.1 - 7 877
FQ210598.1 - 7 877
FQ218289.1 - 8 1077

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001145859, NM_001145858, NM_011893, AK155898, AK150265, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
SH3 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0