This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCL6 SIRNA Assays ›
Gene Symbol:
Bcl6
Gene Name:
B-cell CLL/lymphoma 6
Gene Aliases:
-
Chromosome Location:
Chr.11: 80255722 - 80279077 on Build Rnor_6.0
UniGene ID:
Rn.20571
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS357483
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bcl6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107084.1 NP_001100554.1 8 1943
XM_008768799.1 XP_008767021.1 7 2816
GenBank mRNA BC166425.1 - 8 1843 7097735

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009744, AK036975, U41465, BC052315, AK151229, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
negative regulation of cell-matrix adhesion
germinal center formation
negative regulation of type 2 immune response
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
Rho protein signal transduction
spermatogenesis
protein localization
biological_process
negative regulation of cell proliferation
actin cytoskeleton organization
B cell differentiation
negative regulation of cell growth
positive regulation of B cell proliferation
positive regulation of histone deacetylation
negative regulation of mast cell cytokine production
negative regulation of Rho protein signal transduction
type 2 immune response
regulation of cell proliferation
positive regulation of apoptotic process
negative regulation of apoptotic process
regulation of memory T cell differentiation
negative regulation of cell differentiation
negative regulation of T-helper 2 cell differentiation
positive regulation of neuron differentiation
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
negative regulation of isotype switching to IgE isotypes
erythrocyte development
regulation of inflammatory response
positive regulation of cellular component movement
negative regulation of cellular senescence
RNA polymerase II regulatory region sequence-specific DNA binding
intronic transcription regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
molecular_function
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
chromatin DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0