This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WNT9B SIRNA Assays ›
Gene Symbol:
Wnt9b
Gene Name:
wingless-type MMTV integration site family, member 9B
Gene Aliases:
-
Chromosome Location:
Chr.10: 91785893 - 91807937 on Build Rnor_6.0
UniGene ID:
Rn.16625
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS357409
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wnt9b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107055.1 NP_001100525.1 4 901
XM_006247529.2 XP_006247591.1 4 1711
XM_006247530.2 XP_006247592.1 5 1916

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
in utero embryonic development
kidney development
regulation of protein phosphorylation
regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
biological_process
cellular response to starvation
regulation of asymmetric cell division
Wnt signaling pathway
neuron differentiation
male genitalia development
regulation of tube size
positive regulation of catalytic activity
cell fate commitment
embryonic cranial skeleton morphogenesis
branching morphogenesis of an epithelial tube
palate development
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
kidney morphogenesis
uterus morphogenesis
kidney rudiment formation
mesenchymal stem cell maintenance involved in nephron morphogenesis
collecting duct development
establishment of planar polarity involved in nephron morphogenesis
nephron tubule morphogenesis
mesonephric tubule development
metanephric tubule development
metanephric tubule formation
mesonephric duct formation
molecular_function
frizzled binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0