This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RND2 SIRNA Assays ›
Gene Symbol:
Rnd2
Gene Name:
Rho family GTPase 2
Gene Aliases:
Arhn, RhoN, Rohn
Chromosome Location:
Chr.10: 89376530 - 89380101 on Build Rnor_6.0
UniGene ID:
Rn.22162
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS357385
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010953.1 NP_001010953.1 2 281
GenBank mRNA BC089058.1 - 2 281 7315480
D82867.1 - 2 311

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009708, AF016482, BC139330, BC139331, AK075970, and others.

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
positive regulation of collateral sprouting
GTP binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0