This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMP28 SIRNA Assays ›
Gene Symbol:
Mmp28 LOC103694864
Gene Name:
matrix metallopeptidase 28
matrix metalloproteinase-28-like
Gene Aliases:
-
-
Chromosome Location:
Chr.10: 70660691 - 70681672 on Build Rnor_6.0
Chromosome Location:
Chr.Un|NW_007906801: 53675 - 75488 on Build Rnor_6.0
UniGene ID:
Rn.60472
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS357332
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mmp28
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079888.1 NP_001073357.1 8 1446
GenBank mRNA EF121988.1 - 8 1446
EF121989.1 - 8 1446
Gene Symbol: LOC103694864
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773811.1 XP_008772033.1 8 1637

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
negative regulation of macrophage chemotaxis
metalloendopeptidase activity
calcium ion binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0