This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMGT1 SIRNA Assays ›
Gene Symbol:
Mmgt1
Gene Name:
membrane magnesium transporter 1
Gene Aliases:
RGD1566339, Tmem32
Chromosome Location:
Chr.X: 158966694 - 158978995 on Build Rnor_6.0
UniGene ID:
Rn.64172
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS357163
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mmgt1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106970.1 NP_001100440.1 1, 2 221
GenBank mRNA BC168927.1 - 1, 2 142 7108747

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146234, AK043524, AK163269, AK036253, BC032271, and others.

Gene Ontology Categories:

Function(s) Process(es)

cation transport
cobalt ion transport
copper ion transport
ferrous iron transport
magnesium ion transport
iron ion transmembrane transport
cation transmembrane transport
cobalt ion transmembrane transporter activity
ferrous iron transmembrane transporter activity
magnesium ion transmembrane transporter activity
inorganic cation transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0