This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCL21 SIRNA Assays ›
Gene Symbol:
Ccl21
Gene Name:
chemokine (C-C motif) ligand 21
Gene Aliases:
Ccl21b
Chromosome Location:
Chr.5: 58197678 - 58198782 on Build Rnor_6.0
UniGene ID:
Rn.39658
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS356028
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccl21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008513.1 NP_001008513.1 2 146
GenBank mRNA BC086571.1 - 2 146 7304213
FQ218040.1 - 2 170
FQ220091.1 - 2 170
FQ223815.1 - 2 172
FQ225478.1 - 2 170
FQ226079.1 - 2 168
FQ233622.1 - 2 170

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

establishment of T cell polarity
positive regulation of cell-matrix adhesion
dendritic cell chemotaxis
immune response
G-protein coupled receptor signaling pathway
positive regulation of actin filament polymerization
positive regulation of pseudopodium assembly
ruffle organization
positive regulation of cell adhesion mediated by integrin
response to prostaglandin E
mesangial cell-matrix adhesion
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of protein kinase activity
positive regulation of JNK cascade
release of sequestered calcium ion into cytosol
positive regulation of filopodium assembly
positive regulation of protein kinase B signaling
cell chemotaxis
positive regulation of ERK1 and ERK2 cascade
positive regulation of neutrophil chemotaxis
negative regulation of leukocyte tethering or rolling
negative regulation of leukocyte apoptotic process
positive regulation of cell motility
positive regulation of T cell migration
positive regulation of myeloid dendritic cell chemotaxis
chemokine activity
chemokine receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header