siRNA Details
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039686.1 | NP_001034775.1 | 5 | 379 | ||
XM_008763064.1 | XP_008761286.1 | 4 | 1907 | |||
XM_008763065.1 | XP_008761287.1 | 8 | 863 | |||
GenBank mRNA | AY943907.1 | - | 5 | 379 |
