This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMD5 SIRNA Assays ›
Gene Symbol:
Psmd5
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 13916199 - 13937038 on Build Rnor_6.0
UniGene ID:
Rn.13415
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS355727
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106569.2 NP_001100039.2 4, 5 578
XM_006233969.2 XP_006234031.1 4, 5 1107
XM_006233970.2 XP_006234032.1 5, 6 1197
GenBank mRNA BC166850.1 - 4, 5 560 9088649

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_005047, AK291051, D31889, BC014478, S79862.

Gene Ontology Categories:

Function(s)

proteasome assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0