This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSME2 SIRNA Assays ›
Gene Symbol:
Psme2
Gene Name:
proteasome (prosome, macropain) activator subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 34265552 - 34269851 on Build Rnor_6.0
UniGene ID:
Rn.127779
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS355526
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psme2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017257.2 NP_058953.1 2, 3 88
GenBank mRNA BC058486.1 - 2, 3 126 6918985
BC097331.1 - 2, 3 88 7389180
D45250.1 - 2, 3 138
FQ226805.1 - 2, 3 119
FQ227049.1 - 2, 3 132
FQ232840.1 - 2, 3 119
FQ233763.1 - 2, 3 114

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endopeptidase activity
antigen processing and presentation of exogenous antigen
endopeptidase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0