This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIMK2 SIRNA Assays ›
Gene Symbol:
Limk2
Gene Name:
LIM domain kinase 2
Gene Aliases:
Limk2b, Link2
Chromosome Location:
Chr.14: 83564048 - 83641995 on Build Rnor_6.0
UniGene ID:
Rn.11013
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS355238
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Limk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024135.2 NP_077049.2 15 1730
XM_006251236.2 XP_006251298.1 14 3047
XM_008770275.1 XP_008768497.1 15 1833
XM_008770276.1 XP_008768498.1 14 2864
XM_008770277.1 XP_008768499.1 14 3047
XM_008770278.1 XP_008768500.1 15 1832
XM_008770279.1 XP_008768501.1 15 1832
GenBank mRNA BC065578.1 - 14 1915 5623751
D31874.1 - 15 1730
D31875.1 - 14 1959
D31876.1 - 15 1978
D31877.1 - 15 2102

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
spermatogenesis
spermatid development
protein kinase activity
protein serine/threonine kinase activity
ATP binding
zinc ion binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0