This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HTT SIRNA Assays ›
Gene Symbol:
Htt
Gene Name:
huntingtin
Gene Aliases:
Hd, Hdh
Chromosome Location:
Chr.14: 81105139 - 81254637 on Build Rnor_6.0
UniGene ID:
Rn.11193
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354963
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Htt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024357.3 NP_077333.2 3, 4 576
XM_006251230.2 XP_006251292.1 3, 4 562
XM_006251232.2 XP_006251294.1 3, 4 562
GenBank mRNA AY487897.1 - 3, 4 469
U18650.1 - 3, 4 423

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

urea cycle
citrulline metabolic process
establishment of mitotic spindle orientation
protein import into nucleus
mitochondrial transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
apoptotic process
mitochondrion organization
endoplasmic reticulum organization
Golgi organization
dopamine receptor signaling pathway
spermatogenesis
gastrulation
central nervous system development
brain development
cell aging
learning or memory
learning
grooming behavior
locomotory behavior
axo-dendritic transport
associative learning
determination of adult lifespan
visual learning
anatomical structure morphogenesis
anterior/posterior pattern specification
endosomal transport
lactate biosynthetic process from pyruvate
quinolinate biosynthetic process
striatum development
olfactory lobe development
neural plate formation
neurogenesis
peptide hormone secretion
insulin secretion
positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
regulation of protein phosphatase type 2A activity
social behavior
hormone metabolic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
regulation of mitochondrial membrane permeability
vesicle transport along microtubule
regulation of synaptic plasticity
paraxial mesoderm formation
animal organ development
neuron development
mRNA transport
neuron apoptotic process
response to calcium ion
regulation of mitochondrial membrane potential
L-glutamate import
iron ion homeostasis
negative regulation of neuron death
negative regulation of cysteine-type endopeptidase activity
negative regulation of extrinsic apoptotic signaling pathway
p53 binding
receptor binding
protein binding
transcription factor binding
dynactin binding
identical protein binding
ion channel binding
dynein intermediate chain binding
beta-tubulin binding
diazepam binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0