This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SRPK3 SIRNA Assays ›
Gene Symbol:
Srpk3
Gene Name:
SRSF protein kinase 3
Gene Aliases:
Mssk1, Stk23
Chromosome Location:
Chr.X: 157008773 - 157013455 on Build Rnor_6.0
UniGene ID:
Rn.72618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354815
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Srpk3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_184045.2 NP_908934.1 5 455
XM_006229558.2 XP_006229620.1 5 572
XM_006229559.2 XP_006229621.1 5 571
XM_006229560.2 XP_006229622.1 5 569
XM_006229561.2 XP_006229623.1 5 569
XM_008773617.1 XP_008771839.1 5 569
XM_008773618.1 XP_008771840.1 5 565
GenBank mRNA AY346151.1 - 5 459

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
skeletal muscle tissue development
muscle tissue development
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header