This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PALB2 SIRNA Assays ›
Gene Symbol:
Palb2
Gene Name:
partner and localizer of BRCA2
Gene Aliases:
RGD1304759
Chromosome Location:
Chr.1: 192064586 - 192088547 on Build Rnor_6.0
UniGene ID:
Rn.43638
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354599
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Palb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191603.1 NP_001178532.1 10, 11 2879
XM_008759711.1 XP_008757933.1 8, 9 4055
XM_008759712.1 XP_008757934.1 9, 10 2963
XM_008759713.1 XP_008757935.1 9, 10 2873
GenBank mRNA FQ211178.1 - 10, 11 3028

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081238, BC055302, AK154227, AK157382, BC066140, and others.

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
somitogenesis
inner cell mass cell proliferation
mesoderm development
biological_process
organ morphogenesis
post-anal tail morphogenesis
negative regulation of apoptotic process
molecular_function
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0