This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HTATIP2 SIRNA Assays ›
Gene Symbol:
Htatip2
Gene Name:
HIV-1 Tat interactive protein 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 105094332 - 105109459 on Build Rnor_6.0
UniGene ID:
Rn.138436
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354418
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Htatip2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106263.2 NP_001099733.1 1 183
XM_006229249.2 XP_006229311.1 2 334
XM_006229250.2 XP_006229312.1 2 238
XM_006229251.1 XP_006229313.1 2 249
GenBank mRNA BC158803.1 - 1 183 7321759

Back To Top

More Information


Gene Ontology Categories:

Function(s)

regulation of angiogenesis
nuclear import

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0