This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STMN3 SIRNA Assays ›
Gene Symbol:
Stmn3
Gene Name:
stathmin-like 3
Gene Aliases:
Sclip
Chromosome Location:
Chr.3: 176808016 - 176816108 on Build Rnor_6.0
UniGene ID:
Rn.14961
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354195
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stmn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024346.1 NP_077322.1 3 285
GenBank mRNA AY004290.1 - 3 285
FQ211987.1 - 2 137
FQ212062.1 - 2 146
FQ213831.1 - 2 140
FQ214184.1 - 2 142

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule depolymerization
regulation of microtubule polymerization or depolymerization
cytoplasmic microtubule organization
neuron projection development
negative regulation of Rac protein signal transduction
regulation of cytoskeleton organization
protein binding
protein C-terminus binding
tubulin binding
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0