This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARG1 SIRNA Assays ›
Gene Symbol:
Arg1
Gene Name:
arginase 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 21525421 - 21537872 on Build Rnor_6.0
UniGene ID:
Rn.9857
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS354149
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017134.3 NP_058830.2 8 967
GenBank mRNA BC091158.1 - 8 897 7368991
FQ209619.1 - 5 478
FQ218591.1 - 8 916
FQ219159.1 - 8 849
J02720.1 - 8 848

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007482, U51805, AK149471, BC050005, AK171417, and others.

Gene Ontology Categories:

Function(s) Process(es)

urea cycle
liver development
positive regulation of endothelial cell proliferation
arginine metabolic process
female pregnancy
aging
response to wounding
response to herbicide
response to manganese ion
response to zinc ion
response to selenium ion
regulation of L-arginine import
response to amine
arginine catabolic process to ornithine
lung development
response to lipopolysaccharide
collagen biosynthetic process
response to vitamin A
response to vitamin E
response to drug
response to amino acid
response to peptide hormone
response to cadmium ion
response to steroid hormone
response to axon injury
response to methylmercury
mammary gland involution
maternal process involved in female pregnancy
protein homotrimerization
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
cellular response to interleukin-4
cellular response to glucagon stimulus
cellular response to dexamethasone stimulus
cellular response to transforming growth factor beta stimulus
arginase activity
manganese ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0