siRNA Details
Gene Symbol: Ushbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033687.1 | NP_001028859.1 | 9 | 1513 | ||
GenBank mRNA | BC101905.1 | - | 9 | 1513 | 7934347 |

Gene Symbol: Ushbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033687.1 | NP_001028859.1 | 9 | 1513 | ||
GenBank mRNA | BC101905.1 | - | 9 | 1513 | 7934347 |