This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CENPL SIRNA Assays ›
Gene Symbol:
Cenpl
Gene Name:
centromere protein L
Gene Aliases:
RGD1311868
Chromosome Location:
Chr.13: 78886111 - 78901025 on Build Rnor_6.0
UniGene ID:
Rn.3322
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS353307
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cenpl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033061.2 NP_001028233.2 5 1200
NM_001244761.1 NP_001231690.1 4 1114
XM_006250099.2 XP_006250161.1 5 1252
XM_006250100.2 XP_006250162.1 4 1165
GenBank mRNA BC079467.1 - 5 1200 7122576
BC089893.1 - 4 1114
BC103642.1 - 5 1193 7300924

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header