This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HSBP1 SIRNA Assays ›
Gene Symbol:
Hsbp1
Gene Name:
heat shock factor binding protein 1
Gene Aliases:
Hsp25
Chromosome Location:
Chr.19: 51985196 - 51989528 on Build Rnor_6.0
UniGene ID:
Rn.2578
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS352577
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hsbp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173119.2 NP_775142.1 2, 3 195
GenBank mRNA AF522937.1 - 2, 3 142
BC058131.1 - 2, 3 180 6919748
EX491168.1 - 2, 3 195
FQ211891.1 - 2, 3 163
FQ212621.1 - 2, 3 167
FQ213728.1 - 2, 3 137
FQ216175.1 - 2, 3 174
FQ229155.1 - 2, 3 180
FQ229754.1 - 2, 3 154
FQ230047.1 - 2, 3 152
FQ230722.1 - 2, 3 154
FQ233773.1 - 2, 3 153

Back To Top

More Information


Gene Ontology Categories:

Function(s)

muscle contraction
endodermal cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0