This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TUB SIRNA Assays ›
Gene Symbol:
Tub
Gene Name:
tubby bipartite transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.1: 173550537 - 173632194 on Build Rnor_6.0
UniGene ID:
Rn.30017
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS352275
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tub
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013077.1 NP_037209.1 9 1052
XM_008759690.1 XP_008757912.1 9 1328
XM_008759691.1 XP_008757913.1 8 1161
GenBank mRNA AB011544.1 - 9 1052
U92546.1 - 2 181

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
phagocytosis, recognition
phototransduction
sensory perception of sound
detection of visible light
response to hormone
multicellular organismal macromolecule metabolic process
photoreceptor cell maintenance
positive regulation of phagocytosis
retina development in camera-type eye
receptor localization to nonmotile primary cilium
protein localization to ciliary membrane
molecular_function
G-protein coupled photoreceptor activity
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0