This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ID3 SIRNA Assays ›
Gene Symbol:
Id3
Gene Name:
inhibitor of DNA binding 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 154489603 - 154491172 on Build Rnor_6.0
UniGene ID:
Rn.2760
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS352253
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Id3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013058.2 NP_037190.1 1 94
GenBank mRNA AF000942.1 - 1 87
BC064658.1 - 1 94 5623701
D10864.1 - 1 71
FQ212997.1 - 1 116
FQ216267.1 - 1 116
FQ218468.1 - 1 114
FQ218958.1 - 1 107
FQ229332.1 - 1 116

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
metanephros development
regulation of DNA replication
transcription, DNA-templated
central nervous system development
heart development
muscle organ development
circadian rhythm
positive regulation of cell proliferation
response to wounding
neuron differentiation
epithelial cell differentiation
notochord development
odontogenesis
positive regulation of apoptotic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of cell differentiation
negative regulation of myoblast differentiation
negative regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
regulation of cell cycle
transcription factor activity, sequence-specific DNA binding
transcription factor binding
protein domain specific binding
bHLH transcription factor binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0