This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PNOC SIRNA Assays ›
Gene Symbol:
Pnoc
Gene Name:
prepronociceptin
Gene Aliases:
N23K, Npnc1
Chromosome Location:
Chr.15: 48805842 - 48833053 on Build Rnor_6.0
UniGene ID:
Rn.87935
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS352206
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pnoc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013007.1 NP_037139.1 1, 2 140
XM_006252196.2 XP_006252258.1 2, 3 302
XM_006252199.2 XP_006252261.1 2, 3 300
GenBank mRNA FQ214340.1 - 2, 3 187
FQ220805.1 - 2, 3 189
S79730.1 - 1, 2 77
U48262.1 - 1, 2 143
X97375.1 - 1, 2 140

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neuropeptide signaling pathway
chemical synaptic transmission
opioid peptide activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header