This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GDF7 SIRNA Assays ›
Gene Symbol:
Gdf7
Gene Name:
growth differentiation factor 7
Gene Aliases:
-
Chromosome Location:
Chr.6: 33496596 - 33507626 on Build Rnor_6.0
UniGene ID:
Rn.92437
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351976
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gdf7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170350.1 NP_001163821.1 2 401
XM_006239878.2 XP_006239940.1 2 506

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

axon guidance
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
roof plate formation
spinal cord association neuron differentiation
neural tube development
gland morphogenesis
BMP signaling pathway
epithelial cell differentiation
midbrain development
activin receptor signaling pathway
growth
regulation of apoptotic process
regulation of MAPK cascade
cell fate commitment
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
cell development
reproductive structure development
branching morphogenesis of an epithelial tube
forebrain morphogenesis
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
morphogenesis of an epithelial fold
positive regulation of tendon cell differentiation
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0