This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NRAS SIRNA Assays ›
Gene Symbol:
Nras
Gene Name:
neuroblastoma ras oncogene
Gene Aliases:
-
Chromosome Location:
Chr.2: 205553119 - 205563716 on Build Rnor_6.0
UniGene ID:
Rn.217722
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351462
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nras
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080766.2 NP_542944.1 4 460
XM_006233059.2 XP_006233121.1 4 504
GenBank mRNA BC098659.1 - 4 460 7368145

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010937, BC058755, AK165960, AK010412, CT010359, and others.

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
biological_process
positive regulation of cell proliferation
visual learning
actin cytoskeleton organization
regulation of synaptic transmission, GABAergic
positive regulation of Rac protein signal transduction
negative regulation of neuron apoptotic process
negative regulation of cell differentiation
regulation of long-term neuronal synaptic plasticity
negative regulation of skeletal muscle tissue development
striated muscle cell differentiation
epithelial tube branching involved in lung morphogenesis
molecular_function
protein binding
GTP binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0