This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INHA SIRNA Assays ›
Gene Symbol:
Inha
Gene Name:
inhibin alpha
Gene Aliases:
-
Chromosome Location:
Chr.9: 82700482 - 82703383 on Build Rnor_6.0
UniGene ID:
Rn.8831
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351335
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Inha
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012590.2 NP_036722.1 1 157
GenBank mRNA BC083564.1 - 1 157 7112717

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
male gonad development
positive regulation of pathway-restricted SMAD protein phosphorylation
regulation of cell proliferation
hemoglobin biosynthetic process
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of follicle-stimulating hormone secretion
cell development
regulation of cell cycle
SMAD protein signal transduction
receptor binding
cytokine activity
transforming growth factor beta receptor binding
hormone activity
growth factor activity
inhibin binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0