This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GLS2 SIRNA Assays ›
Gene Symbol:
Gls2
Gene Name:
glutaminase 2 (liver, mitochondrial)
Gene Aliases:
Ga
Chromosome Location:
Chr.7: 2605693 - 2621853 on Build Rnor_6.0
UniGene ID:
Rn.10202
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351098
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gls2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270786.1 NP_001257715.1 6 977
NM_001270787.1 NP_001257716.1 6 1223
NM_138904.2 NP_620259.2 5 672
XM_006240728.2 XP_006240790.1 6 1003
XM_006240730.2 XP_006240792.1 7 802
GenBank mRNA BC089776.1 - 6 816
BC104712.1 - 6 721
FQ211735.1 - 6 1223
J05499.1 - 5 671

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: AF279698.
This siRNA also targets the following transcripts in Mouse: NM_001285779, NM_001285777, NM_001033264, EU770627, AK220231, and others.

Gene Ontology Categories:

Function(s) Process(es)

glutamine metabolic process
regulation of apoptotic process
reactive oxygen species metabolic process
glutaminase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0