This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRLF2 SIRNA Assays ›
Gene Symbol:
Crlf2
Gene Name:
cytokine receptor-like factor 2
Gene Aliases:
Crl2, Tpte2, Tslpr
Chromosome Location:
Chr.14: 1456843 - 1461543 on Build Rnor_6.0
UniGene ID:
Rn.177948
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351004
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crlf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134465.2 NP_604460.2 4 555
XM_008769915.1 XP_008768137.1 4 513
XM_008769916.1 XP_008768138.1 4 513
GenBank mRNA AF404510.1 - 4 498
BC078740.1 - 4 555 7108137
FQ232217.1 - 4 541

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immunoglobulin secretion involved in immune response
inflammatory response
T-helper 2 cell cytokine production
cytokine activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0