This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NLRP6 SIRNA Assays ›
Gene Symbol:
Nlrp6 LOC103690037
Gene Name:
NLR family, pyrin domain containing 6
NACHT, LRR and PYD domains-containing protein 6-like
Gene Aliases:
Avr, Nalp6, Navr, Navr/Avr
-
Chromosome Location:
Chr.1: 213686017 - 213692177 on Build Rnor_6.0
Chromosome Location:
Chr.1: 213676328 - 213683616 on Build Rnor_6.0
UniGene ID:
Rn.9669
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS350937
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nlrp6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134375.3 NP_599202.2 1536
GenBank mRNA DQ631800.1 - 1536
M85183.1 - 412
Gene Symbol: LOC103690037
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006230507.2 XP_006230569.2 3 2697

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133946, BC031139, FJ222825, BC013519, JF810536, and others.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
response to bacterium
regulation of autophagy
negative regulation of toll-like receptor signaling pathway
wound healing
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAPK cascade
regulation of inflammatory response
negative regulation of immune response
regulation of mucus secretion
vasopressin receptor activity
ATP binding
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header